Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: itr-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027999.4 | NP_001023170.1 | 9 - 10 | 1336 | 122 | |
NM_001028000.5 | NP_001023171.1 | 7 - 8 | 1168 | 122 | ||
NM_001028002.4 | NP_001023173.1 | 8 - 9 | 1264 | 122 | ||
NM_001268376.4 | NP_001255305.1 | 8 - 9 | 1198 | 122 | ||
NM_001380339.1 | NP_001367466.1 | 8 - 9 | 1198 | 122 | ||
GenBank mRNA | AF168688.1 | - | 8 - 9 | 1264 | 122 | |
AJ243179.2 | - | 8 - 9 | 1286 | 122 | ||
AJ243180.2 | - | 9 - 10 | 1359 | 122 | ||
AJ243181.2 | - | 8 - 9 | 1279 | 122 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |