Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ceh-19 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027970.4 | NP_001023141.1 | 3 - 4 | 229 | 153 | |
NM_001027971.6 | NP_001023142.1 | 3 - 4 | 580 | 153 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |