Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-24 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001368443.4 | NP_001355386.1 | 1 - 2 | 235 | 128 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |