Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: hyl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307101.3 | NP_001294030.1 | 4 - 5 | 430 | 114 | |
NM_001380373.1 | NP_001368453.1 | 3 - 4 | 409 | 114 | ||
NM_069058.9 | NP_501459.1 | 3 - 4 | 418 | 114 | ||
GenBank mRNA | AF105010.1 | - | 3 - 4 | 418 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Recommended Assay Caenorhabditis elegans Probe spans exons |