Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: twk-8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028425.5 | NP_001023596.1 | 5 - 6 | 1110 | 73 | |
NM_001028426.5 | NP_001023597.1 | 5 - 6 | 1116 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |