Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069190.4 | NP_501591.2 | 9 - 10 | 1798 | 172 | |
GenBank mRNA | DQ360810.1 | - | 9 - 10 | 1797 | 172 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |