Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306928.3 | NP_001293857.1 | 2 - 3 | 184 | 80 | |
NM_001306929.3 | NP_001293858.1 | 2 - 3 | 191 | 80 | ||
NM_001380409.1 | NP_001368188.1 | 3 - 4 | 249 | 80 | ||
NM_001380410.1 | NP_001366749.1 | 3 - 4 | 249 | 80 | ||
NM_069252.7 | NP_501653.1 | 2 - 3 | 191 | 80 | ||
GenBank mRNA | AY396716.1 | - | 3 - 4 | 249 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |