Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: daf-15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306928.3 | NP_001293857.1 | 11 - 12 | 3305 | 93 | |
NM_001380409.1 | NP_001368188.1 | 12 - 13 | 3361 | 93 | ||
NM_001380410.1 | NP_001366749.1 | 12 - 13 | 3361 | 93 | ||
GenBank mRNA | AY396716.1 | - | 12 - 13 | 3361 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |