Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: amph-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069310.5 | NP_501711.1 | 8 - 9 | 842 | 114 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |