Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069374.10 | NP_501775.1 | 2 - 3 | 180 | 93 | |
GenBank mRNA | AF083223.1 | - | 1 - 2 | 97 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |