Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tbc-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001027993.4 | NP_001023164.1 | 4 - 5 | 561 | 72 | |
NM_001027994.4 | NP_001023165.1 | 5 - 6 | 634 | 72 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |