Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069421.6 | NP_501822.1 | 2 - 3 | 293 | 108 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |