Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cwn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069421.6 | NP_501822.1 | 5 - 6 | 977 | 75 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |