Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gst-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069446.4 | NP_501847.1 | 2 - 3 | 287 | 89 | |
GenBank mRNA | AF010240.1 | - | 1 - 2 | 229 | 89 |
Gene Symbol: gst-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069445.7 | NP_501846.1 | 1 - 2 | 255 | 89 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |