Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gst-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069447.7 | NP_501848.1 | 3 - 4 | 455 | 80 | |
GenBank mRNA | AF010239.1 | - | 3 - 4 | 350 | 80 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Inventoried Probe spans exons |