Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: abt-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306925.2 | NP_001293854.1 | 16 - 17 | 2552 | 162 | |
NM_001306926.3 | NP_001293855.1 | 13 - 14 | 2315 | 162 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |