Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ech-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380437.1 | NP_001368477.1 | 5 - 6 | 856 | 98 | |
NM_069474.6 | NP_501875.2 | 5 - 6 | 911 | 98 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |