Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: gpa-7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069520.3 | NP_501921.1 | 4 - 5 | 732 | 176 | |
GenBank mRNA | AY008130.1 | - | 4 - 5 | 721 | 176 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |