Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tax-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026121.2 | NP_001021292.1 | 4 - 5 | 478 | 67 | |
NM_001083189.5 | NP_001076658.2 | 4 - 5 | 478 | 67 | ||
NM_001380448.1 | NP_001367052.1 | 3 - 4 | 226 | 67 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |