Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cyb-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069586.8 | NP_501987.1 | 5 - 6 | 1020 | 104 | |
GenBank mRNA | U20903.1 | - | 4 - 5 | 991 | 104 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |