Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dnc-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268592.3 | NP_001255521.1 | 9 - 10 | 1770 | 97 | |
NM_001268593.3 | NP_001255522.1 | 3 - 4 | 883 | 97 | ||
NM_001276852.4 | NP_001263781.1 | 7 - 8 | 1568 | 97 | ||
NM_001276853.3 | NP_001263782.1 | 8 - 9 | 1686 | 97 | ||
NM_001383200.1 | 9 - 10 | 1815 | 97 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |