Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-41 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307551.3 | NP_001294480.1 | 7 - 8 | 1412 | 87 | |
NM_001307552.3 | NP_001294481.1 | 6 - 7 | 1310 | 87 | ||
NM_001307576.3 | NP_001294505.1 | 6 - 7 | 1364 | 87 | ||
NM_001307577.3 | NP_001294506.1 | 5 - 6 | 1244 | 87 | ||
NM_067672.5 | NP_500073.3 | 9 - 10 | 1690 | 87 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Recommended Assay Caenorhabditis elegans Probe spans exons |