Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: ubxn-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001028419.5 | NP_001023590.1 | 4 - 5 | 671 | 73 | |
NM_001380105.1 | NP_001368066.1 | 2 - 3 | 347 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Inventoried Probe spans exons |