Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: mca-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268315.3 | NP_001255244.1 | 10 - 11 | 1331 | 79 | |
NM_001307544.3 | NP_001294473.1 | 4 - 5 | 388 | 79 | ||
NM_001383061.1 | 9 - 10 | 1228 | 79 | |||
NM_001383062.1 | 9 - 10 | 1230 | 79 | |||
NM_001383063.1 | 9 - 10 | 1230 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |