Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: cal-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307362.2 | NP_001294291.1 | 3 - 4 | 280 | 80 | |
NM_068020.7 | NP_500421.2 | 4 - 5 | 481 | 80 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |