Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-72 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068041.6 | NP_500442.3 | 5 - 6 | 1012 | 150 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |