Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: clec-78 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068053.5 | NP_500454.2 | 14 - 15 | 4515 | 81 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |