Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: oac-52 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068145.3 | NP_500546.2 | 9 - 10 | 1548 | 79 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |