Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: wapl-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380190.1 | NP_001368333.1 | 2 - 3 | 225 | 93 | |
NM_068165.6 | NP_500566.1 | 2 - 3 | 225 | 93 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |