Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fbxb-77 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307467.4 | NP_001294396.1 | 1 - 2 | 98 | 68 | |
NM_068233.6 | NP_500634.2 | 1 - 2 | 113 | 68 |
Set Membership: |
Amplicon length less than or equal to 70 3' Most Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |