Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eor-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380207.2 | NP_001367921.1 | 14 - 15 | 2489 | 90 | |
NM_001380208.1 | NP_001368235.1 | 3 - 4 | 624 | 90 | ||
GenBank mRNA | AF502568.1 | - | 15 - 16 | 2597 | 90 | |
AF519108.1 | - | 14 - 15 | 2478 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |