Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: rme-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001380231.1 | NP_001368360.1 | 5 - 6 | 2427 | 90 | |
NM_068414.6 | NP_500815.2 | 6 - 7 | 2732 | 90 |
Set Membership: |
Amplicon length between 86 and 100 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |