Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: glf-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068483.5 | NP_500884.2 | 2 - 3 | 416 | 152 | |
GenBank mRNA | AJ871144.2 | - | 2 - 3 | 415 | 152 |
Set Membership: |
Amplicon length greater than or equal to 101 Caenorhabditis elegans Probe spans exons |