Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-44 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001026096.4 | NP_001021267.1 | 1 - 2 | 94 | 71 | |
NM_001380249.1 | NP_001367345.1 | 9 - 10 | 3154 | 71 | ||
NM_001380250.2 | NP_001367343.1 | 8 - 9 | 3155 | 71 | ||
NM_001380251.2 | NP_001367344.1 | 8 - 9 | 3152 | 71 | ||
NM_001380252.2 | NP_001368407.1 | 9 - 10 | 3159 | 71 | ||
NM_001380253.1 | NP_001368408.1 | 3 - 4 | 637 | 71 | ||
NM_001380254.1 | NP_001368409.1 | 4 - 5 | 655 | 71 | ||
NM_068500.5 | NP_500901.2 | 2 - 3 | 424 | 71 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |