Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: pqn-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306867.3 | NP_001293796.1 | 11 - 12 | 1987 | 90 | |
NM_001306871.3 | NP_001293800.1 | 10 - 11 | 1849 | 90 | ||
NM_001306878.3 | NP_001293807.1 | 11 - 12 | 2104 | 90 | ||
NM_068506.3 | NP_500907.2 | 12 - 13 | 2242 | 90 | ||
NM_068507.6 | NP_500908.2 | 4 - 5 | 592 | 90 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |