Hamburger Menu Button
Thermo Fisher Scientific Logo
Sign in
Don't have an account ?
Create Account
Products
Antibodies
Cell Culture Media
Chemicals
Chromatography Columns and Cartridges
Lab Equipment
Lab Plasticware and Supplies
Microplates
Oligos, Primers, Probes and Genes
TaqMan Real-Time PCR Assays
Greener Products
See all product categories
Applications
Bioprocessing
Cell Culture and Transfection
Cell and Gene Therapy
Chromatography
Molecular Testing
Digital Solutions
DNA and RNA Extraction and Analysis
Spectroscopy, Elemental and Isotope Analysis
See all applications and techniques
Services
Integrated CRO-CDMO Services
CDMO Services
CRO Services
Custom Services
Enterprise Services
Financial and Leasing Services
Instrument Services
Lab Informatics
OEM and Commercial Supply
Training Services
Unity Lab Services
See all services
Help and Support
How to Order
Instrument Support
Learning Centers
Register for an Account
Technical Support Centers
See all help and support topics
Popular
TaqMan Real-Time PCR Assays
Antibodies
Oligos, Primers & Probes
GeneArt Gene Synthesis
Cell Culture Plastics
Contact Us
Quick Order
Order Status and Tracking
Documents and Certificates
Thermo Fisher Scientific Logo
Search
Thermo Fisher Scientific
Search All
Search
Search button
Search button
Close
Order Status
Quick Order
Sign in
Sign in
Don't have an account ?
Create Account
Account
Check Order Status
Aspire Member Program
Connect: Lab, Data, Apps
Custom Products & Projects
Services Central
This product has been added to your favorites list. Go to
My Favorites
System Message
OK
Cancel
LOADING
Home
›
TaqMan
®
Gene Expression Assays
›
Search Tool
›
Search Results
› Ce02464277_g1
See other PQN-22 GE Assays ›
Gene Symbol:
pqn-22
Gene Name:
protein-coding
Gene Aliases:
CELE_C46G7.4
Chromosome Location:
Chr.IV: 6019672 - 6033639 on Build WBcel235
Species:
Caenorhabditis elegans
Species Specific ID (Flybase ID):
-
Assay ID
Ce02464277_g1
Size
FAM-MGB | XS: 75 rxns
FAM-MGB | S: 360 rxns
FAM-MGB | M: 750 rxns
FAM-MGB | L: 2900 rxns
VIC-MGB_PL | S: 360 rxns
VIC-MGB_PL | M: 750 rxns
VIC-MGB_PL | L: 2900 rxns
VIC-MGB | S: 360 rxns
VIC-MGB | M: 750 rxns
VIC-MGB | L: 2900 rxns
Availability
Made To Order
Catalog #
4351372
Price
Your Price
Online offer:
Check your price ›
Add To Cart
Genomic Map
Assay Details
More Information
Genomic Map
LOADING...
×
Back To Top
Assay Details
Gene Symbol: pqn-22
Interrogated Sequence
Translated Protein
Exon Boundary
Assay Location
IMAGE Clone ID
Amplicon Length
RefSeq
NM_001306867.3
NP_001293796.1
17 - 18
2629
106
NM_001306868.2
NP_001293797.1
16 - 17
2572
106
NM_001306869.4
NP_001293798.1
14 - 15
2365
106
NM_001306870.4
NP_001293799.1
13 - 14
2302
106
NM_001306871.3
NP_001293800.1
16 - 17
2491
106
NM_001306872.3
NP_001293801.1
15 - 16
2434
106
NM_001306873.3
NP_001293802.1
13 - 14
2227
106
NM_001306874.3
NP_001293803.1
13 - 14
2419
106
NM_001306875.3
NP_001293804.1
17 - 18
2827
106
NM_001306876.3
NP_001293805.1
15 - 16
2620
106
NM_001306877.4
NP_001293806.1
14 - 15
2557
106
NM_001306878.3
NP_001293807.1
17 - 18
2746
106
NM_001306879.3
NP_001293808.1
16 - 17
2689
106
NM_001306880.3
NP_001293809.1
14 - 15
2482
106
NM_001380255.1
NP_001368420.1
6 - 7
907
106
NM_001380256.2
NP_001368419.1
7 - 8
970
106
NM_001380257.1
NP_001368418.1
9 - 10
1177
106
NM_068505.4
NP_500906.2
12 - 13
2164
106
NM_068506.3
NP_500907.2
18 - 19
2884
106
NM_068507.6
NP_500908.2
10 - 11
1234
106
Back To Top
More Information
Set Membership:
Amplicon length greater than or equal to 101
Detect Genomic DNA
Caenorhabditis elegans
Probe spans exons
Back To Top
Related Products
TaqMan
®
Fast Advanced Master Mix
SuperScript
®
VILO
™
cDNA Synthesis Kit
AddToFavs result goes here(DO NOT DELETE ME!)
Close X
Your item
s have
has
been added!
Host server : magellan-search-7487cff77c-wzvj9:80/10.239.161.253:80.
git-commit: c531a7a49d92b854cb004d8acd820c41186dcba6
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.16.2-2024.09.38-1.0