Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-31 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068550.5 | NP_500951.2 | 4 - 5 | 744 | 113 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |