Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: srv-32 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_068554.2 | NP_500955.1 | 7 - 8 | 899 | 90 |
Set Membership: |
Amplicon length between 86 and 100 G-protein coupled receptor 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |