Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: best-9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001383107.1 | 5 - 6 | 580 | 119 |
Gene Symbol: try-3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001306808.3 | NP_001293737.1 | 4 - 5 | 528 | 119 | |
NM_001380280.2 | NP_001367393.1 | 5 - 6 | 753 | 119 |
Set Membership: |
Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |