Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: tag-89 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069746.2 | NP_502147.1 | 3 - 4 | 482 | 131 |
Set Membership: |
Amplicon length greater than or equal to 101 G-protein coupled receptor Detect Genomic DNA Caenorhabditis elegans Probe spans exons |