Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: kin-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268622.2 | NP_001255551.1 | 16 - 17 | 3521 | 75 | |
NM_001268624.3 | NP_001255553.1 | 16 - 17 | 3512 | 75 | ||
NM_001268625.3 | NP_001255554.1 | 15 - 16 | 2975 | 75 | ||
NM_001268626.3 | NP_001255555.1 | 14 - 15 | 2963 | 75 | ||
NM_001268627.2 | NP_001255556.1 | 16 - 17 | 3512 | 75 | ||
NM_001268629.3 | NP_001255558.1 | 11 - 12 | 2174 | 75 | ||
NM_001307048.3 | NP_001293977.1 | 13 - 14 | 2732 | 75 | ||
NM_001380461.1 | NP_001367231.1 | 15 - 16 | 3416 | 75 | ||
NM_001380462.2 | NP_001366913.1 | 15 - 16 | 3455 | 75 | ||
GenBank mRNA | AM086627.1 | - | 13 - 14 | 2723 | 75 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |