Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: magi-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268648.3 | NP_001255577.1 | 13 - 14 | 3092 | 72 | |
NM_001268650.4 | NP_001255579.1 | 8 - 9 | 2682 | 72 | ||
NM_001268652.3 | NP_001255581.1 | 6 - 7 | 2366 | 72 | ||
NM_001383205.1 | 14 - 15 | 3192 | 72 | |||
NM_001383206.1 | 13 - 14 | 3098 | 72 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |