Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lips-5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001083237.3 | NP_001076706.1 | 2 - 3 | 289 | 73 | |
NM_001268659.2 | NP_001255588.1 | 2 - 3 | 395 | 73 | ||
NM_001268660.3 | NP_001255589.1 | 2 - 3 | 292 | 73 |
Set Membership: |
Amplicon length between 71 and 85 Caenorhabditis elegans Probe spans exons |