Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307047.3 | NP_001293976.1 | 5 - 6 | 742 | 87 | |
NM_069853.5 | NP_502254.1 | 5 - 6 | 677 | 87 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |