Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: nhr-43 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001307047.3 | NP_001293976.1 | 6 - 7 | 1146 | 106 | |
NM_069853.5 | NP_502254.1 | 6 - 7 | 1081 | 106 |
Set Membership: |
Amplicon length greater than or equal to 101 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |