Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: fat-6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001268666.3 | NP_001255595.1 | 3 - 4 | 289 | 66 | |
NM_001268667.3 | NP_001255596.1 | 1 - 2 | 104 | 66 | ||
GenBank mRNA | AF260244.1 | - | 2 - 3 | 236 | 66 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |