Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: unc-22 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129363.4 | NP_001122835.2 | 15 - 16 | 5947 | 94 | |
NM_001268672.3 | NP_001255601.1 | 21 - 22 | 6610 | 94 | ||
NM_001268673.3 | NP_001255602.1 | 22 - 23 | 6373 | 94 | ||
NM_001268674.4 | NP_001255603.2 | 12 - 13 | 5185 | 94 | ||
NM_001268675.3 | NP_001255604.1 | 11 - 12 | 5134 | 94 | ||
NM_001307045.3 | NP_001293974.1 | 24 - 25 | 7255 | 94 | ||
NM_001307046.3 | NP_001293975.1 | 23 - 24 | 7228 | 94 | ||
NM_069872.6 | NP_502273.2 | 21 - 22 | 6473 | 94 | ||
NM_069873.6 | NP_502274.2 | 24 - 25 | 7303 | 94 |
Set Membership: |
Amplicon length between 86 and 100 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |