Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: lex-1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001129296.3 | NP_001122768.1 | 1 - 2 | 129 | 65 | |
NM_069888.9 | NP_502289.2 | 1 - 2 | 129 | 65 | ||
GenBank mRNA | DQ140399.1 | - | 1 - 2 | 127 | 65 |
Set Membership: |
Amplicon length less than or equal to 70 Detect Genomic DNA Recommended Assay Caenorhabditis elegans Probe spans exons |