Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: dpy-26 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069977.8 | NP_502378.1 | 8 - 9 | 3016 | 83 | |
GenBank mRNA | U43562.1 | - | 8 - 9 | 2971 | 83 |
Set Membership: |
Amplicon length between 71 and 85 Detect Genomic DNA Caenorhabditis elegans Probe spans exons |