Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: eak-4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_069997.2 | NP_502398.1 | 2 - 3 | 409 | 73 | |
GenBank mRNA | DQ907008.1 | - | 2 - 3 | 409 | 73 |
Set Membership: |
Amplicon length between 71 and 85 3' Most Detect Genomic DNA Caenorhabditis elegans Probe spans exons |